MilliporeSigma
All Photos(1)

ATE959437979

AstaTech, Inc.

DICHLOROBIS(TRIETHYLPHOSPHINE)PALLADIUM(II)

CAS Number:
Molecular Weight:
415.66
MDL number:

Iupac Name

bis(triethyl-lambda5-phosphaneyl)palladium(IV) chloride

Inchi Code

1S/2C6H15P.2ClH.Pd/c2*1-4-7(5-2)6-3;;;/h2*4-6H2,1-3H3;2*1H;

InChI key

YQLNDFXCSZLJNP-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=109312

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F16634

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service