Skip to Content
MilliporeSigma

ATE959438316

AstaTech, Inc.

CHLOROBIS(ETHYLENE)RHODIUM(I) DIMER

Synonym(s):

bis(λ¹-rhodium(1+)) tetrakis(ethene) dichloride

Sign In to View Organizational & Contract Pricing.

Select a Size


Inchi Code

1S/4C2H4.2ClH.2Rh/c4*1-2;;;;/h4*1-2H2;2*1H;;/q;;;;;;2*+1/p-2

Inchi Key

OOQJCPOXJFCGCR-UHFFFAOYSA-L


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library