Documents

ATEH1F1C5641

AstaTech, Inc.

(S)-METHYL 2-(N-(2,6-DIMETHYLPHENYL)-2-PHENYLACETAMIDO)PROPANOATE

CAS Number:
Molecular Weight:
325.41
MDL number:

Iupac Name

methyl N-(2,6-dimethylphenyl)-N-(2-phenylacetyl)-L-alaninate

Inchi Code

1S/C20H23NO3/c1-14-9-8-10-15(2)19(14)21(16(3)20(23)24-4)18(22)13-17-11-6-5-7-12-17/h5-12,16H,13H2,1-4H3/t16-/m0/s1

InChI key

CJPQIRJHIZUAQP-INIZCTEOSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=132431

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=A50836

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.