Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

ATEH3031F010

AstaTech, Inc.

2-METHYLUNDECANE

Synonym(s):

2-methylundecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ATEH3031F010 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
170.34
MDL number:

Skip To

Purity

80%

Inchi Code

1S/C12H26/c1-4-5-6-7-8-9-10-11-12(2)3/h12H,4-11H2,1-3H3

Inchi Key

GTJOHISYCKPIMT-UHFFFAOYSA-N

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library