Skip to Content
MilliporeSigma

ATEH75DBF98A

AstaTech, Inc.

AZIDO-PEG6-ACID

Sign In to View Organizational & Contract Pricing

Select a Size

100 MG

$23.75

250 MG

$47.50

1 G

$143.75

Vendor SKU: C70161-100MG

$23.75


Ships in 10 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
379.41
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

1-azido-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid

Inchi Code

1S/C15H29N3O8/c16-18-17-2-4-22-6-8-24-10-12-26-14-13-25-11-9-23-7-5-21-3-1-15(19)20/h1-14H2,(H,19,20)

InChI key

KQYQHDQBMAJDRL-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://astatechinc.com/CPSResult.php?CRNO=147652

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service