Skip to Content
MilliporeSigma

ATEH96F19BE8

AstaTech, Inc.

4-NITROCINNAMYL ALCOHOL

Synonym(s):

(2E)-3-(4-nitrophenyl)-2-propen-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H9O3N1
CAS Number:
Molecular Weight:
179.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H9NO3/c11-7-1-2-8-3-5-9(6-4-8)10(12)13/h1-6,11H,7H2/b2-1+

Inchi Key

LGXXEDSIJZHDBN-OWOJBTEDSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

2-8 C

Country of Origin

CN

Physical Form

Solid



Signal Word

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library