All Photos(1)

ATEH97E805B6

AstaTech, Inc.

MZ 1

CAS Number:
Molecular Weight:
1002.66
MDL number:

Iupac Name

(2S,4R)-1-((S)-2-(tert-butyl)-17-((S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-4,16-dioxo-6,9,12-trioxa-3,15-diazaheptadecanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Inchi Code

1S/C49H60ClN9O8S2/c1-28-30(3)69-48-41(28)42(33-12-14-35(50)15-13-33)54-37(45-57-56-31(4)59(45)48)23-39(61)51-16-17-65-18-19-66-20-21-67-26-40(62)55-44(49(5,6)7)47(64)58-25-36(60)22-38(58)46(63)52-24-32-8-10-34(11-9-32)43-29(2)53-27-68-43/h8-15,27,36-38,44,60H,16-26H2,1-7H3,(H,51,61)(H,52,63)(H,55,62)/t36-,37+,38+,44-/m1/s1

InChI key

PTAMRJLIOCHJMQ-PYNGZGNASA-N

storage temp.

2-8 C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=191887

Signal Word

Warning

Hazard Statements

H302;H315;H319;H335

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AT14161

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service