Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

ATEH991806BD

AstaTech, Inc.

AFIMOXIFENE

Synonym(s):

4-((1E)-1-{4-[2-(dimethylamino)ethoxy]phenyl}-2-phenyl-1-butenyl)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ATEH991806BD is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
387.52
MDL number:

Skip To

Inchi Code

1S/C26H29NO2/c1-4-25(20-8-6-5-7-9-20)26(21-10-14-23(28)15-11-21)22-12-16-24(17-13-22)29-19-18-27(2)3/h5-17,28H,4,18-19H2,1-3H3/b26-25+

Inchi Key

TXUZVZSFRXZGTL-OCEACIFDSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library