Skip to Content
MilliporeSigma

ATEH996C557B

AstaTech, Inc.

2-IODO-2-METHYLPROPANE

Synonym(s):

2-iodo-2-methylpropane

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ATEH996C557B is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
184.02
MDL number:

Skip To

Inchi Code

1S/C4H9I/c1-4(2,3)5/h1-3H3

Inchi Key

ANGGPYSFTXVERY-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Liquid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library