Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

ATEH99996732

AstaTech, Inc.

RESAZURIN SODIUM SALT

Synonym(s):

sodium 10-oxo-10H-phenoxazin-10-ium-3,7-bis(olate)

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
251.17
MDL number:

Inchi Code

1S/C12H7NO4.Na/c14-7-1-3-9-11(5-7)17-12-6-8(15)2-4-10(12)13(9)16;/h1-6H,(H-,14,15,16);/q;+1/p-1

Inchi Key

OPVDVVJXMPZWLP-UHFFFAOYSA-M

Purity

95%

Country of Origin

CN

Physical Form

Light-green to Brown Solid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library