Skip to Content
MilliporeSigma

ATEH9A7F0644

AstaTech, Inc.

METHOXY-PEG-MALEIMIDE

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product ATEH9A7F0644 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
243.26
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

1-(2-(2-(2-methoxyethoxy)ethoxy)ethyl)-1H-pyrrole-2,5-dione

Inchi Code

1S/C11H17NO5/c1-15-6-7-17-9-8-16-5-4-12-10(13)2-3-11(12)14/h2-3H,4-9H2,1H3

InChI key

WQUWKZJWBCOHQH-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

https://astatechinc.com/CPSResult.php?CRNO=268329

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library