Skip to Content
MilliporeSigma

ATEH9ACDDE9F

AstaTech, Inc.

DICHLORO(PENTAMETHYLCYCLOPENTADIENYL)IRIDIUM(III) DIMER

Synonym(s):

iridium(III) bis(1,2,3,4,5-pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ATEH9ACDDE9F is currently not available for purchase. Contact Technical Service

Skip To

Shipping Temperature

Normal

Storage temp.

2-8 C

Inchi Code

1S/2C10H15.4ClH.2Ir/c2*1-6-7(2)9(4)10(5)8(6)3;;;;;;/h2*1-5H3;4*1H;;/q2*-1;;;;;2*+3/p-4

Inchi Key

MMAGMBCAIFVRGJ-UHFFFAOYSA-J

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library