Skip to Content
MilliporeSigma

ATEH9ADF6F3D

AstaTech, Inc.

ACID RED 14

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product ATEH9ADF6F3D is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
502.44
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

sodium (E)-4-(2-(1-oxo-4-sulfonatonaphthalen-2(1H)-ylidene)hydrazineyl)naphthalene-1-sulfonate

Inchi Code

1S/C20H14N2O7S2.2Na/c23-20-15-8-4-3-7-14(15)19(31(27,28)29)11-17(20)22-21-16-9-10-18(30(24,25)26)13-6-2-1-5-12(13)16;;/h1-11,21H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b22-17+;;

InChI key

OJNIRNOPCQQNRY-JZFXEGLSSA-L

Purity

95%

Country of Origin

CN

Physical Form

Yellow to Brown Solid

Product Link

https://astatechinc.com/CPSResult.php?CRNO=210981

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library