MilliporeSigma
All Photos(1)

ATC359510213

Atlantic Research Chemicals Ltd

Ethyl 2-Methylcyclopropanecarboxylate

CAS Number:
Molecular Weight:
128.17
MDL number:

Iupac Name

ethyl 2-methylcyclopropanecarboxylate

Inchi Code

1S/C7H12O2/c1-3-9-7(8)6-4-5(6)2/h5-6H,3-4H2,1-2H3

InChI key

JWYSLVLBKXDZCW-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=ES09902

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service