Skip to Content
MilliporeSigma

ATCH99D3AFC9

Atlantic Research Chemicals Ltd

2-Chloro-1,3-dimethylimidazolinium chloride

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
171.07
MDL number:

Iupac Name

2-chloro-1,3-dimethylimidazolidin-1-ium chloride

Inchi Code

1S/C5H11ClN2.ClH/c1-7-3-4-8(2)5(7)6;/h5H,3-4H2,1-2H3;1H

InChI key

BNGPVKSKKYIJCR-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=HA01019

MSDS

https://atlantic-chemicals.com/product/?code=HA01019


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library