Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATCH99D3B5FA

Atlantic Research Chemicals Ltd

Methanol, sodium salt (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
55.03
MDL number:
Product ATCH99D3B5FA is currently not available for purchase. Contact Technical Service

Iupac Name

methanol, sodium salt

Inchi Code

1S/CH4O.Na/c1-2;/h2H,1H3;

InChI key

YWOITFUKFOYODT-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=AN047043

MSDS

https://atlantic-chemicals.com/product/?code=AN047043


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library