MilliporeSigma
All Photos(2)

700008P

Avanti

15-ketocholestene

Avanti Polar Lipids 700008P, powder

Synonym(s):
3β-hydroxy-5α-cholest-8(14)-en-15-one
Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700008P-5mg)

manufacturer/tradename

Avanti Polar Lipids 700008P

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C27H44O2/c1-17(2)7-6-8-18(3)23-16-24(29)25-21-10-9-19-15-20(28)11-13-26(19,4)22(21)12-14-27(23,25)5/h17-20,22-23,28H,6-16H2,1-5H3/t18?,19-,20-,22?,23+,26-,27+/m0/s1

InChI key

LINVVMHRTUSXHL-NDNSGUFDSA-N

General description

15-ketocholestene is the oxidized derivative of cholesterol and is associated with plasma.

Application

15-ketocholestene has been used to test its binding to RAb2E9 recombinant monoclonal (RAb) antibody using enzyme-linked immunosorbent assay (ELISA). It has also been used as an antigen to test its reactivity with IgG in multiple sclerosis samples.

Packaging

5 mL Amber Glass Screw Cap Vial (700008P-5mg)

Storage Class Code

13 - Non Combustible Solids

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service