MilliporeSigma
All Photos(2)

700010P

Avanti

15α-hydroxycholestene

5α-cholest-8(14)-ene-3β,15α-diol, powder

Synonym(s):
3b,15a-diol-5a-cholest-8(14)-ene
Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700010P-5mg)

manufacturer/tradename

Avanti Polar Lipids 700010P

shipped in

dry ice

storage temp.

−20°C

General description

15α-hydroxycholestene is the oxidized derivative of cholesterol.

Biochem/physiol Actions

15α-hydroxycholestene levels are modulated in the relapsing-remitting multiple sclerosis (RRMS) and secondary progressive multiple sclerosis (SPMS). It is implicated in autoimmune encephalomyelitis (EAE).

Packaging

5 mL Amber Glass Screw Cap Vial (700010P-5mg)

Storage Class Code

13 - Non Combustible Solids

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service