MilliporeSigma
All Photos(2)

700011P

Avanti

15β-hydroxycholestene

5α-cholest-8(14)-ene-3β,15β,-diol, powder

Synonym(s):
3b,15b-diol-5a-cholest-8(14)-ene
Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700011P-5mg)

manufacturer/tradename

Avanti Polar Lipids 700011P

shipped in

dry ice

storage temp.

−70°C

General description

15β-hydroxycholestene is a derivative of cholesterol.

Application

15β-hydroxycholestene has been used in a study to identify specific antibodies against lipids using phage display.

Packaging

5 mL Amber Glass Screw Cap Vial (700011P-5mg)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Generation and characterization of a novel recombinant antibody against 15-ketocholestane isolated by phage-display
Islam MD, et al.
International Journal of Molecular Sciences, 13(4), 4937-4948 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service