Skip to Content
MilliporeSigma

BL3H1F1CB96F

BLD Pharmatech Co., Ltd.

4-Cyano-3-fluorophenyl 4-propylbenzoate

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product BL3H1F1CB96F is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
283.3
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

4-cyano-3-fluorophenyl 4-propylbenzoate

Inchi Code

1S/C17H14FNO2/c1-2-3-12-4-6-13(7-5-12)17(20)21-15-9-8-14(11-19)16(18)10-15/h4-10H,2-3H2,1H3

Inchi Key

RDPCRVRASQOZBX-UHFFFAOYSA-N

storage temp.

Sealed in dry, room temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Form: Crystal - Powder / Colour: White - Almost white

Product Link

http://www.bldpharm.com/products/86776-51-4.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.bldpharm.com/static/upload/prosds/bldsds/138/SDS-BD137291.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library