A-125

Supelco

Multi-component Alcohol Mix

2000 μg/mL each component in H2O, certified reference material, Cerilliant®

Quality Level

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Acetone
Ethanol
Isopropanol
Methanol

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

2000 μg/mL each component in H2O

format

multi-component solution

storage temp.

2-8°C

InChI

1S/C15H13N3O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9,16H2,(H,18,19)/i1D,2D,3D,4D,5D

InChI key

OYOUQHVDCKOOAL-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A multicomponent calibration kit containing acetone, ethanol, isopropanol, and methanol at a concentration of 2000 μg/mL. This kit is suitable for calibration of blood alcohol testing methods using GC Headspace.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Cerilliant Corporation

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.