Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-164

Supelco

7-Hydroxywarfarin solution

1 mg/mL (Degassed Acetonitrile with 0.1% BHT), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

4,7-Dihydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one, 7-Hydroxywarfarin, Coumarin, 3-(α-acetonylbenzyl)-4,7-dihydroxy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16O5
CAS Number:
Molecular Weight:
324.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Degassed Acetonitrile with 0.1% BHT)

format

single component solution

storage temp.

-10 to -25°C

SMILES string

O1c2c(ccc(c2)O)C(=O)C(C1=O)C(CC(=O)C)c3ccccc3

InChI key

JAGKHISTOCDVMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F

flash_point_c

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service