All Photos(1)

CH4182077102

ChemBridge Corporation

3-amino-4-methylbenzenesulfonamide

CAS Number:
Molecular Weight:
186.23
MDL number:

Iupac Name

3-amino-4-methylbenzenesulfonamide

Inchi Code

1S/C7H10N2O2S/c1-5-2-3-6(4-7(5)8)12(9,10)11/h2-4H,8H2,1H3,(H2,9,10,11)

InChI key

HCZDHXMDYWZPPN-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

RU

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/4028971

MSDS

https://www.hit2lead.com/sds/4028971_sds.pdf

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service