MilliporeSigma
All Photos(1)

CH6371386340

Chem Impex International, Inc.

Wieland-Miescher ketone

CAS Number:
Molecular Weight:
178.23
MDL number:

mp

47-50

Iupac Name

8a-methyl-3,4,8,8a-tetrahydro-1,6(2H,7H)-naphthalenedione

Inchi Code

1S/C11H14O2/c1-11-6-5-9(12)7-8(11)3-2-4-10(11)13/h7H,2-6H2,1H3

InChI key

DNHDRUMZDHWHKG-UHFFFAOYSA-N

storage temp.

Store at 0-8

Purity

99%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Beige to yellowish crystalline powder

Product Link

https://www.chemimpex.com/product/productinfo/plusmn-8a-methyl-3-4-8-8a-tetrahydro-1-6-2-i-h-i-7-i-h-i-naphthalenedione/14661?cid=0

Melting Point

47-50

MSDS

https://www.chemimpex.com/product/productinfo/plusmn-8a-methyl-3-4-8-8a-tetrahydro-1-6-2-i-h-i-7-i-h-i-naphthalenedione/14661?cid=0

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service