All Photos(1)

CH6371386382

Chem Impex International, Inc.

1-(p-Hydroxyphenyl)ethylamine

CAS Number:
Molecular Weight:
137.18
MDL number:

Iupac Name

4-(1-aminoethyl)phenol

Inchi Code

1S/C8H11NO/c1-6(9)7-2-4-8(10)5-3-7/h2-6,10H,9H2,1H3

InChI key

CDQPLIAKRDYOCB-UHFFFAOYSA-N

storage temp.

Store at 0-8

Purity

99%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Light tan solid

Product Link

https://www.chemimpex.com/product/productinfo/1-i-pi-i-hydroxyphenyl-ethylamine/14704?cid=0

MSDS

https://www.chemimpex.com/product/productinfo/1-i-pi-i-hydroxyphenyl-ethylamine/14704?cid=0

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service