Select a Size
$24.08
$61.06
$172.51
$323.66
$1,114.07
Vendor SKU: 00531-5G
$24.08
About This Item
Skip To
Iupac Name
3,6-anhydro-4-O-[3-O-(3,6-anhydro-4-O-beta-D-galactopyranosyl-alpha-L-galactopyranosyl)-beta-D-galactopyranosyl]-alpha-L-galactopyranose
Inchi Code
1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1
InChI key
MJQHZNBUODTQTK-WKGBVCLCSA-N
storage temp.
Country of Origin
Shipping Temperature
Product Link
MSDS
https://www.chemimpex.com/products/00531
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service