All Photos(1)

CH6H11E4EF54

Chem Impex International, Inc.

DL-Adrenaline

CAS Number:
Molecular Weight:
183.21
MDL number:

mp

197 �dec)

Iupac Name

4-[1-hydroxy-2-(methylamino)ethyl]-1,2-benzenediol

Inchi Code

1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3

InChI key

UCTWMZQNUQWSLP-UHFFFAOYSA-N

storage temp.

Store at 2-8

Purity

98%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Pale yellow crystalline powder

Product Link

https://www.chemimpex.com/product/productinfo/dl-adrenaline/21655?cid=1

Melting Point

197 �dec)

MSDS

https://www.chemimpex.com/product/productinfo/dl-adrenaline/21655?cid=1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service