CH6H97ACC4EF

Chem Impex International, Inc.

Chitosan , low molecular weight (20-300 cP, 1 wt. % in 1% acetic acid at 25 �- 50,000-190,000 Da

CAS Number:
Molecular Weight:
161.16
MDL number:

Iupac Name

3-amino-2,6-anhydro-1,3-dideoxy-5-O-methyl-D-glycero-D-gulo-heptitol

Purity

96.5%

Country of Origin

US

Product Link

https://www.chemimpex.com/product/productinfo/chitosan-low-molecular-weight-20-300-cp-1-wt-in-1-acetic-acid-at-25-deg-c-50-000-190-000-da-based-on-viscosity-/26128?cid=1

MSDS

https://www.chemimpex.com/product/productinfo/chitosan-low-molecular-weight-20-300-cp-1-wt-in-1-acetic-acid-at-25-deg-c-50-000-190-000-da-based-on-viscosity-/26128?cid=1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service