Skip to Content
MilliporeSigma

CH6H9A56B201

Chem Impex International, Inc.

Inosine

Synonym(s):

2-(6-hydroxy-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CH6H9A56B201 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C10H12O5N4
CAS Number:
Molecular Weight:
268.23
MDL number:

Skip To

Inchi Code

1S/C10H12N4O5/c15-1-4-6(16)7(17)10(19-4)14-3-13-5-8(14)11-2-12-9(5)18/h2-4,6-7,10,15-17H,1H2,(H,11,12,18)

Inchi Key

UGQMRVRMYYASKQ-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

99%

Storage temp.

Store at RT

Country of Origin

US

Physical Form

White crystalline powder

Melting Point

212-213C (dec)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library