Skip to Content
MilliporeSigma

CH6H9A56B23A

Chem Impex International, Inc.

Sodium sulfate, anhydrous

Synonym(s):

sulfuric acid, sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

1 KG

$32.50

2.5 KG

$65.00

10 KG

$175.50

25 KG

$370.50

Vendor SKU: 00274-1KG

$32.50


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
121.07
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

density

= 88 lbs/ft3

Inchi Code

1S/Na.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)

Inchi Key

PANBYUAFMMOFOV-UHFFFAOYSA-N

Storage temp.

Store at RT

Purity

99%

Country of Origin

US

Shipping Temperature

RT

MSDS

https://www.chemimpex.com/products/00274


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service