Skip to Content
MilliporeSigma

CH6H9A56B2C5

Chem Impex International, Inc.

5-Bromo-4-chloro-3-indolyl-b-D-glucuronide sodium salt

Synonym(s):

6-((5-bromo-4-chloro-1H-indol-3-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid, sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

100 MG

$45.50

250 MG

$84.50

1 G

$266.50

5 G

$1,118.00

Vendor SKU: 00631-100MG

$45.50


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
445.61
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C14H13BrClNO7.Na/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;/h1-3,9-12,14,17-20H,(H,21,22);

Inchi Key

LGUWKXOPXWJLQX-UHFFFAOYSA-N

Storage temp.

Store at 0-8C

Purity

98%

Country of Origin

US

Shipping Temperature

RT

MSDS

https://www.chemimpex.com/products/00631


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service