Skip to Content
MilliporeSigma

CH6H9A56B2DE

Chem Impex International, Inc.

Na-Fmoc-Nim-trityl-L-histidine

Synonym(s):

N-[(9H-fluoren-9-ylmethoxy)carbonyl]-1-tritylhistidine

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$26.00

25 G

$45.50

100 G

$143.00

250 G

$338.00

1 KG

$1,287.00

Vendor SKU: 00713-5G

$26.00


Ships in 7 days - from Aldrich Partner.


About This Item

Linear Formula:
C40H33O4N3
CAS Number:
Molecular Weight:
619.72
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C40H33N3O4/c44-38(45)37(42-39(46)47-26-36-34-22-12-10-20-32(34)33-21-11-13-23-35(33)36)24-31-25-43(27-41-31)40(28-14-4-1-5-15-28,29-16-6-2-7-17-29)30-18-8-3-9-19-30/h1-23,25,27,36-37H,24,26H2,(H,42,46)(H,44,45)

Inchi Key

XXMYDXUIZKNHDT-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

99.7%99%

Storage temp.

Store at 0 - 8 C

Country of Origin

US

Physical Form

White powder or crystals

Melting Point

134 - 153 C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service