Skip to Content
MilliporeSigma

CH6H9A56B346

Chem Impex International, Inc.

Bromophenol blue sodium salt

Synonym(s):

sodium 3,5-dibromo-4-(3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CH6H9A56B346 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
691.95
MDL number:

Skip To

Inchi Code

1S/C19H10Br4O5S.Na/c20-12-5-9(24)6-13(21)17(12)19(18-14(22)7-10(25)8-15(18)23)11-3-1-2-4-16(11)29(26,27)28-19;/h1-8,24-25H;/q;+1/p-1

Inchi Key

KGANHHDZDYZJEM-UHFFFAOYSA-M

Storage temp.

Store at RT

Country of Origin

US

Shipping Temperature

RT

MSDS

https://www.chemimpex.com/products/01224


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library