Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6H9A56B359

Chem Impex International, Inc.

Potassium sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$24.05
250 G
$45.50
1 KG
$91.00
2.5 KG
$175.50
5 KG
$312.00
10 KG
$578.50
25 KG
$1,332.50

Vendor SKU: 01254-100G

$24.05


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 G
$24.05
250 G
$45.50
1 KG
$91.00
2.5 KG
$175.50
5 KG
$312.00
10 KG
$578.50
25 KG
$1,332.50

About This Item

CAS Number:
Molecular Weight:
137.18
MDL number:

Vendor SKU: 01254-100G

$24.05


Ships in 7 days - from Aldrich Partner.

mp

1069 C (Approx.)

Iupac Name

sulfuric acid, potassium salt

Inchi Code

1S/K.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)

InChI key

ILRLTAZWFOQHRT-UHFFFAOYSA-N

storage temp.

Store at RT

Purity

99%

Country of Origin

US

Shipping Temperature

RT

Physical Form

White crystals

Product Link

https://www.chemimpex.com/product/productinfo/potassium-sulfate/775?cid=0

Melting Point

1069 C (Approx.)

MSDS

https://www.chemimpex.com/product/productinfo/potassium-sulfate/775?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service