Skip to Content
MilliporeSigma

CH6H9A56B36B

Chem Impex International, Inc.

Adenosine-5'-triphosphate disodium salt trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$26.00
25 G
$39.00
100 G
$117.00
250 G
$253.50
1 KG
$799.50

Vendor SKU: 00015-5G

$26.00


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
605.19
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

176C (dec)

Iupac Name

sodium (5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen triphosphate trihydrate

Inchi Code

1S/C10H16N5O13P3.2Na.3H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;3*1H2/q;2*+1;;;/p-2

InChI key

MWEQTWJABOLLOS-UHFFFAOYSA-L

storage temp.

Store at RT

Purity

98%

Country of Origin

US

Shipping Temperature

RT

Product Link

https://www.chemimpex.com/products/00015

Melting Point

176C (dec)

MSDS

https://www.chemimpex.com/products/00015


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service