Skip to Content
MilliporeSigma

CH6H9A56B3C2

Chem Impex International, Inc.

Choline dihydrogencitrate salt

Sign In to View Organizational & Contract Pricing

Select a Size

Product CH6H9A56B3C2 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
295.29
MDL number:

Skip To

mp

103.0 - 107.5 C

Iupac Name

2-hydroxy-N,N,N-trimethylethanaminium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate

Inchi Code

1S/C6H8O7.C5H14NO/c7-3(8)1-6(13,5(11)12)2-4(9)10;1-6(2,3)4-5-7/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);7H,4-5H2,1-3H3/q;+1/p-1

InChI key

WRPUOFKIGGWQIJ-UHFFFAOYSA-M

storage temp.

Store at 0 - 8 C

Purity

99%

Country of Origin

US

Shipping Temperature

RT

Product Link

https://www.chemimpex.com/products/01440

Melting Point

103.0 - 107.5 C

MSDS

https://www.chemimpex.com/products/01440


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library