Skip to Content
MilliporeSigma

CH6H9A56B42C

Chem Impex International, Inc.

Resazurin sodium salt

Synonym(s):

7-hydroxy-3-oxo-3H-phenoxazine 10-oxide, sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CH6H9A56B42C is currently not available for purchase. Contact Technical Service

Skip To

Inchi Code

1S/C12H7NO4.Na/c14-7-1-3-9-11(5-7)17-12-6-8(15)2-4-10(12)13(9)16;/h1-6,14H;

Inchi Key

UYXSVQYIDVRWQT-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

90%

Storage temp.

Store at RT

Country of Origin

US

Physical Form

Purple-Green-black powder


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library