Skip to Content
MilliporeSigma

CH6H9A56B43C

Chem Impex International, Inc.

Sodium thiosulfate pentahydrate

Sign In to View Organizational & Contract Pricing

Select a Size

250 G

$24.08

1 KG

$41.66

2.5 KG

$88.27

5 KG

$153.76

10 KG

$249.58

25 KG

$567.77

50 KG

$1,080.22

Vendor SKU: 01802-250G

$24.08


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
155.15
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

48 C (Approx.)

Iupac Name

sulfurothioic O,S-acid, sodium salt hydrate

Inchi Code

1S/Na.H2O3S2.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2

InChI key

HBLWERCELPPGQF-UHFFFAOYSA-N

storage temp.

Store at RT

Purity

99.5-101.0%

Country of Origin

US

Shipping Temperature

RT

Product Link

https://www.chemimpex.com/products/01802

Melting Point

48 C (Approx.)

MSDS

https://www.chemimpex.com/products/01802


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service