Sign In to View Organizational & Contract Pricing.
Select a Size
1 G
$24.08
5 G
$54.54
25 G
$219.76
100 G
$697.58
250 G
$1,436.96
Vendor SKU: 04035-1G
$24.08
Ships in 7 days - from Aldrich Partner.
Skip To
Inchi Code
1S/C28H30N2O5/c1-27(2,3)35-26(34)29-23(25(32)33)19-24(31)30-28(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22/h4-18,23H,19H2,1-3H3,(H,29,34)(H,30,31)(H,32,33)
Inchi Key
PYGOCFDOBSXROC-UHFFFAOYSA-N
Shipping Temperature
RT
Purity
98%
Storage temp.
Store at 0-8 C
Country of Origin
US
Physical Form
White powder
Melting Point
180-200 C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service