Skip to Content
MilliporeSigma

CH6H9A56BFD5

Chem Impex International, Inc.

N,N'-bis-Boc-1-guanylpyrazole

Synonym(s):

(E)-tert-butyl (1H-pyrazol-1-yl)methanediylidenedicarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size


Inchi Code

1S/C14H22N4O4/c1-13(2,3)21-11(19)16-10(18-9-7-8-15-18)17-12(20)22-14(4,5)6/h7-9H,1-6H3,(H,16,17,19,20)

Inchi Key

QFNFDHNZVTWZED-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

97%

Storage temp.

Store at 0-8 C

Country of Origin

US

Physical Form

White powder

Melting Point

86-90 C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library