Skip to Content
MilliporeSigma

CH6H9A56C012

Chem Impex International, Inc.

Ne-Acetyl-L-lysine

Synonym(s):

N~6~-acetyllysine

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C8H16O3N2
CAS Number:
Molecular Weight:
188.23
MDL number:

Inchi Code

1S/C8H16N2O3/c1-6(11)10-5-3-2-4-7(9)8(12)13/h7H,2-5,9H2,1H3,(H,10,11)(H,12,13)

Inchi Key

DTERQYGMUDWYAZ-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

98%

Storage temp.

Store at ≤ -4 C

Country of Origin

US

Physical Form

White to off-white powder

Melting Point

220 - 240 C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library