Skip to Content
MilliporeSigma

CH6H9A56D4DA

Chem Impex International, Inc.

6-Chloro-3-indolyl-b-D-galactopyranoside

Synonym(s):

2-((6-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Sign In to View Organizational & Contract Pricing.

Select a Size


Inchi Code

1S/C14H16ClNO6/c15-6-1-2-7-8(3-6)16-4-9(7)21-14-13(20)12(19)11(18)10(5-17)22-14/h1-4,10-14,16-20H,5H2

Inchi Key

OQWBAXBVBGNSPW-UHFFFAOYSA-N

Shipping Temperature

RT

Purity

98%

Storage temp.

Store at 0-8 C

Country of Origin

US

Physical Form

White to off-white cream powder


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library