Skip to Content
MilliporeSigma

CH6H9A56D53B

Chem Impex International, Inc.

g-Cyclodextrin

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$24.08
25 G
$40.62
100 G
$96.08
250 G
$164.82
1 KG
$572.85
5 KG
$2,582.55

Vendor SKU: 21825-5G

$24.08


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
1297.14
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol

Inchi Code

1S/C48H80O40/c49-1-9-33-17(57)25(65)41(73-9)82-34-10(2-50)75-43(27(67)19(34)59)84-36-12(4-52)77-45(29(69)21(36)61)86-38-14(6-54)79-47(31(71)23(38)63)88-40-16(8-56)80-48(32(72)24(40)64)87-39-15(7-55)78-46(30(70)22(39)62)85-37-13(5-53)76-44(28(68)20(37)60)83-35-11(3-51)74-42(81-33)26(66)18(35)58/h9-72H,1-8H2

InChI key

GDSRMADSINPKSL-UHFFFAOYSA-N

storage temp.

Store at 0-8C

Purity

98-102%

Country of Origin

US

Shipping Temperature

RT

Product Link

https://www.chemimpex.com/products/21825

MSDS

https://www.chemimpex.com/products/21825


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service