Skip to Content
MilliporeSigma

CH6H9A56D5EC

Chem Impex International, Inc.

Methyl Red free acid ACS

Synonym(s):

2-{(E)-[4-(dimethylamino)phenyl]diazenyl}benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CH6H9A56D5EC is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
269.3
MDL number:

Skip To

mp

177-184 C

Inchi Code

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)

Inchi Key

CEQFOVLGLXCDCX-UHFFFAOYSA-N

Storage temp.

Store at RT

Purity

96%

Country of Origin

US

Shipping Temperature

RT

MSDS

https://www.chemimpex.com/products/22782


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library