Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6H9A56D8AF

Chem Impex International, Inc.

Indocyanine Green

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
774.98
MDL number:

Iupac Name

sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trienyl)-1,1-dimethyl-1H-benzo[e]indolium-3-yl)butane-1-sulfonate

Inchi Code

1S/C43H48N2O6S2.Na/c1-42(2)38(44(28-14-16-30-52(46,47)48)36-26-24-32-18-10-12-20-34(32)40(36)42)22-8-6-5-7-9-23-39-43(3,4)41-35-21-13-11-19-33(35)25-27-37(41)45(39)29-15-17-31-53(49,50)51;/h5-13,18-27H,14-17,28-31H2,1-4H3,(H-,46,47,48,49,50,51);/q;+1/p-1

InChI key

MOFVSTNWEDAEEK-UHFFFAOYSA-M

storage temp.

Store at 0-8 C

Purity

96%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Dark green to brown crystalline powder

Product Link

https://www.chemimpex.com/product/productinfo/indocyanine-green-nbsp/13462?cid=0

MSDS

https://www.chemimpex.com/product/productinfo/indocyanine-green-nbsp/13462?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library