Skip to Content
MilliporeSigma

CH6H9A56DA69

Chem Impex International, Inc.

trans-3-(4-Methoxybenzoyl)acrylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

Product CH6H9A56DA69 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
206.2
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

mp

128-138 C

Iupac Name

(2E)-4-(4-methoxyphenyl)-4-oxo-2-butenoic acid

Inchi Code

1S/C11H10O4/c1-15-9-4-2-8(3-5-9)10(12)6-7-11(13)14/h2-7H,1H3,(H,13,14)

InChI key

WORYXBDHTBWLLL-UHFFFAOYSA-N

storage temp.

Store at 0-8C

Purity

98%

Country of Origin

US

Shipping Temperature

RT

Product Link

https://www.chemimpex.com/products/25698

Melting Point

128-138 C

MSDS

https://www.chemimpex.com/products/25698


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library