Skip to Content
MilliporeSigma

CH6H9A56DB82

Chem Impex International, Inc.

Etoposide

Sign Into View Organizational & Contract Pricing

Select a Size

Product CH6H9A56DB82 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
588.57
MDL number:

Skip To

Iupac Name

9-(4-hydroxy-3,5-dimethoxyphenyl)-8-oxo-5,5a,6,8,8a,9-hexahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-5-yl 4,6-O-ethylidenehexopyranoside

Inchi Code

1S/C29H32O13/c1-11-36-9-20-27(40-11)24(31)25(32)29(41-20)42-26-14-7-17-16(38-10-39-17)6-13(14)21(22-15(26)8-37-28(22)33)12-4-18(34-2)23(30)19(5-12)35-3/h4-7,11,15,20-22,24-27,29-32H,8-10H2,1-3H3

InChI key

VJJPUSNTGOMMGY-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

95-105%

Country of Origin

US

Shipping Temperature

RT

Product Link

https://www.chemimpex.com/products/28435

MSDS

https://www.chemimpex.com/products/28435


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library