Inchi Code
1S/C28H34O15/c1-10-21(33)23(35)25(37)27(39-10)43-26-24(36)22(34)19(9-29)42-28(26)40-12-6-14(31)20-15(32)8-17(41-18(20)7-12)11-3-4-16(38-2)13(30)5-11/h3-7,10,17,19,21-31,33-37H,8-9H2,1-2H3
Inchi Key
ARGKVCXINMKCAZ-UHFFFAOYSA-N
Shipping Temperature
RT
Purity
98%
Storage temp.
Store at 0-8 C
Country of Origin
US
Physical Form
Light yellow to off-white crystalline powder
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.