Sign In to View Organizational & Contract Pricing.
Select a Size
Product CH6H9A56DF29 is currently not available for purchase. Contact Technical Service
About This Item
Skip To
Inchi Code
1S/C21H22N2O2S/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17/h2-15,20-21,23H,22H2,1H3
Inchi Key
UOPFIWYXBIHPIP-UHFFFAOYSA-N
Shipping Temperature
RT
Purity
99%
Storage temp.
Store at 0-8 C
Country of Origin
US
Physical Form
White to pale yellow crystalline powder
Melting Point
124-129 C
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.