Sign In to View Organizational & Contract Pricing.
Select a Size
Product CH6H9A56DF37 is currently not available for purchase. Contact Technical Service
Skip To
Inchi Code
1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10,19H,2-8,11-18H2,1H3
Inchi Key
ALSTYHKOOCGGFT-UHFFFAOYSA-N
Shipping Temperature
RT
Purity
99%
Storage temp.
Store at 0-8 C
Country of Origin
US
Physical Form
Colorless liquid
Density
0.849 g/mL at 25 C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.